Product Details

SNP ID
rs13476418
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:33218338 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAGACTCCAGCAAGCCTGGCCGTGG[A/T]AACTACTGCCACACAACACCAGCGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Angptl2 PubMed Links

Gene Details

Gene
Angptl2
Gene Name
angiopoietin-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_011923.4 Intron NP_036053.2
Gene
Ralgps1
Gene Name
Ral GEF with PH domain and SH3 binding motif 1
There are no transcripts associated with this gene.

View Full Product Details