Product Details

SNP ID
rs13476945
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:181234960 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGCAGTCTGTGCCTTGGCTGCTGCC[C/T]CGGCTCTGGTCTCTGCCTTCACCGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Helz2 PubMed Links

Gene Details

Gene
Helz2
Gene Name
helicase with zinc finger 2, transcriptional coactivator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183162.2 3875 Missense Mutation NP_898985.2

View Full Product Details