Product Details

SNP ID
rs13471817
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:63151632 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGTACCATGTTGGACACAGCT[A/G]CAAGAATTGCTGCTCCATCATCTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ino80dos PubMed Links

Gene Details

Gene
Ino80dos
Gene Name
INO80 complex subunit D, opposite strand
There are no transcripts associated with this gene.

Gene
Ndufs1
Gene Name
NADH dehydrogenase (ubiquinone) Fe-S protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160038.1 1818 Missense Mutation NP_001153510.1
NM_001160039.1 1818 Missense Mutation NP_001153511.1
NM_001160040.1 1818 Missense Mutation NP_001153512.1
NM_145518.2 1818 Missense Mutation NP_663493.2

View Full Product Details