Product Details

SNP ID
rs13483149
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:86990367 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGTCGTTTAAAAGTAAGGGCTGGTA[C/T]GTGAAACTCTCACGTGTTCTGTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Pigf PubMed Links

Gene Details

Gene
Pigf
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class F
There are no transcripts associated with this gene.

Gene
Rhoq
Gene Name
ras homolog gene family, member Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145491.2 Intron NP_663466.2

View Full Product Details