Product Details

SNP ID
rs3666286
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:58575655 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTTTGCTTTTGTTTCTCTTGC[A/C]CTCCCCTTCTCTGGTCACTCCAGTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Slc27a6 PubMed Links

Gene Details

Gene
Slc27a6
Gene Name
solute carrier family 27 (fatty acid transporter), member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081072.1 Intron NP_001074541.1

View Full Product Details