Product Details
- SNP ID
-
rs13480655
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:75409978 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGTAACAGTGTTTCAGTTTAATC[A/C]TTTAACGCACTCATGACAGAGATAC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Upb1
PubMed Links
Gene Details
- Gene
- Upb1
- Gene Name
- ureidopropionase, beta
There are no transcripts associated with this gene.
View Full Product Details