Product Details

SNP ID
rs13483327
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:42933486 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCCCTGCCCAACACCCCTGGC[C/T]TCCCCTGCAAACATATAATCTGGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
4933407I08Rik PubMed Links

Gene Details

Gene
4933407I08Rik
Gene Name
RIKEN cDNA 4933407I08 gene
There are no transcripts associated with this gene.

Gene
Ppp2r2b
Gene Name
protein phosphatase 2, regulatory subunit B, beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_028392.3 Intron NP_082668.1

View Full Product Details