Product Details

SNP ID
rs13477568
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:13837845 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCTTTAGCAACGGGCCTTCTTCTTC[C/T]TCTTCCTCTTCTCTGGCTAATCAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Runx1t1 PubMed Links

Gene Details

Gene
Runx1t1
Gene Name
runt-related transcription factor 1; translocated to, 1 (cyclin D-related)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111026.2 652 Silent Mutation NP_001104496.1
NM_001111027.2 652 Silent Mutation NP_001104497.1
NM_009822.3 652 Silent Mutation NP_033952.1

View Full Product Details