Product Details

SNP ID
rs6359590
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:83070900 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAGTGGTGTTCCATGCAGCATGTTG[C/T]ATAGTTTAGACATCTTTGCATTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Plrg1 PubMed Links

Gene Details

Gene
Plrg1
Gene Name
pleiotropic regulator 1, PRL1 homolog (Arabidopsis)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016784.3 Intron NP_058064.2

View Full Product Details