Product Details

SNP ID
rs13476393
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:26910617 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCTCCCGCCGCCTCTCGGAGCCTGC[G/T]GTCCGGTGCCAGGGCCCGCCCGCCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Med22 PubMed Links

Gene Details

Gene
Med22
Gene Name
mediator complex subunit 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033908.1 26 UTR 5 NP_001029080.1
NM_011513.2 26 UTR 5 NP_035643.1
Gene
Rpl7a
Gene Name
ribosomal protein L7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013721.3 26 Intron NP_038749.1
Gene
Surf1
Gene Name
surfeit gene 1
There are no transcripts associated with this gene.

Gene
Surf2
Gene Name
surfeit gene 2
There are no transcripts associated with this gene.

Gene
Surf4
Gene Name
surfeit gene 4
There are no transcripts associated with this gene.

Gene
Surf6
Gene Name
surfeit gene 6
There are no transcripts associated with this gene.

View Full Product Details