Product Details
- SNP ID
-
rs13478663
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:24451353 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTTTCCAGCGAGAAATCGGCTTC[C/T]GTCCTTATTTTTGTCCCACTGTATA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Iqub
PubMed Links
Gene Details
- Gene
- Iqub
- Gene Name
- IQ motif and ubiquitin domain containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172535.3 |
|
Intron |
|
|
NP_766123.2 |
View Full Product Details