Product Details
- SNP ID
-
rs13483306
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:37690719 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGTGTCACCCTTCGGGAGGATGT[A/G]CCACCAGGCTTCTCCGTGCTTCAGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Gm37013
PubMed Links
Gene Details
- Gene
- Gm37013
- Gene Name
- predicted gene, 37013
There are no transcripts associated with this gene.
- Gene
- Gm38666
- Gene Name
- predicted gene, 38666
There are no transcripts associated with this gene.
- Gene
- Pcdha7-g
- Gene Name
- protocadherin alpha 7-gamma
There are no transcripts associated with this gene.
- Gene
- Pcdhga1
- Gene Name
- protocadherin gamma subfamily A, 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033584.1 |
861 |
Intron |
|
|
NP_291062.1 |
- Gene
- Pcdhga2
- Gene Name
- protocadherin gamma subfamily A, 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033585.1 |
861 |
Intron |
|
|
NP_291063.1 |
- Gene
- Pcdhga3
- Gene Name
- protocadherin gamma subfamily A, 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033586.2 |
861 |
Intron |
|
|
NP_291064.1 |
- Gene
- Pcdhga4
- Gene Name
- protocadherin gamma subfamily A, 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033587.3 |
861 |
Intron |
|
|
NP_291065.3 |
- Gene
- Pcdhga5
- Gene Name
- protocadherin gamma subfamily A, 5
There are no transcripts associated with this gene.
- Gene
- Pcdhgb1
- Gene Name
- protocadherin gamma subfamily B, 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033574.3 |
861 |
Intron |
|
|
NP_291052.3 |
- Gene
- Pcdhgb2
- Gene Name
- protocadherin gamma subfamily B, 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033575.3 |
861 |
Silent Mutation |
|
|
NP_291053.1 |
View Full Product Details