Product Details

SNP ID
rs13480752
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:106830087 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTTGTTTGTGTGTCACCACAGCATA[A/G]TGAAGGAATTGAACAGGAGAGGCTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ppfia2 PubMed Links

Gene Details

Gene
Ppfia2
Gene Name
protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205341.1 Intron NP_001192270.1
NM_177373.4 Intron NP_796347.2

View Full Product Details