Product Details

SNP ID
rs13481959
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:96666144 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGTACTTGGACCCAAGCTGCCGTAG[A/G]TTCTGGAACTGGAAGTAAATGTACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Hmgcr PubMed Links

Gene Details

Gene
Hmgcr
Gene Name
3-hydroxy-3-methylglutaryl-Coenzyme A reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008255.2 395 Silent Mutation NP_032281.2

View Full Product Details