Product Details

SNP ID
rs6384437
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:104162098 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGCTTTGCACAGGACTCAGATTTGC[G/T]CACCGTGAGGCCAAGTGGAGCTAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Crhr1 PubMed Links

Gene Details

Gene
Crhr1
Gene Name
corticotropin releasing hormone receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007762.4 Intron NP_031788.1

View Full Product Details