Product Details
- SNP ID
-
rs13477109
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:51966439 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTGGTCAGGCAGTAAAAGGACTGT[C/T]GGGATGCTATCTCATTCAACTGGGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Maml3
PubMed Links
Gene Details
- Gene
- Maml3
- Gene Name
- mastermind like 3 (Drosophila)
View Full Product Details