Product Details
- SNP ID
-
rs13482641
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:71917280 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCCTTCTGAGTATTTGCAGGTTC[A/G]GGACAGTCCTGCCAAGGGCCTTTCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Col22a1
PubMed Links
Gene Details
- Gene
- Col22a1
- Gene Name
- collagen, type XXII, alpha 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_027174.1 |
|
Intron |
|
|
NP_081450.1 |
View Full Product Details