Product Details

SNP ID
rs13476503
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:53183609 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCTAAATCCATGTTTTCAAAAGCAA[C/T]GGAAGGTTCTGAAGATGAGGACCTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Arl6ip6 PubMed Links

Gene Details

Gene
Arl6ip6
Gene Name
ADP-ribosylation factor-like 6 interacting protein 6
There are no transcripts associated with this gene.

Gene
Prpf40a
Gene Name
PRP40 pre-mRNA processing factor 40 homolog A (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018785.2 Intron NP_061255.1

View Full Product Details