Product Details
- SNP ID
-
rs4207009
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:77107995 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTCTACTTTATCCAGAACCAGGC[A/G]CCAAAGAAAATCATTGAGAAAACAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Usp25
PubMed Links
Gene Details
- Gene
- Usp25
- Gene Name
- ubiquitin specific peptidase 25
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013918.2 |
2475 |
Silent Mutation |
|
|
NP_038946.2 |
View Full Product Details