Product Details
- SNP ID
-
rs13459053
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:140105459 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATTGAGCGTTCGTACTCCTGCTT[A/G]TCTCTACATTTGTAATCCATGGTGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cfh
PubMed Links
Gene Details
- Gene
- Cfh
- Gene Name
- complement component factor h
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_009888.3 |
2535 |
Silent Mutation |
|
|
NP_034018.2 |
View Full Product Details