Product Details

SNP ID
rs8258353
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:86300892 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTGGGCTTGTCGTGCTCCTGAGCTG[C/T]TGGAGCCTTGGGCACTGGGGCGCGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Syn3 PubMed Links

Gene Details

Gene
Syn3
Gene Name
synapsin III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164495.1 481 Intron NP_001157967.1
NM_013722.3 481 Intron NP_038750.2
Gene
Timp3
Gene Name
tissue inhibitor of metalloproteinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_011595.2 481 Silent Mutation NP_035725.1

View Full Product Details