Product Details
- SNP ID
-
rs13481384
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:33190499 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCAAGTAGCTGTTAATAAAAGTG[C/T]TAGAAGGATGGTAGCAATAATCACA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Atxn7l1
PubMed Links
Gene Details
- Gene
- Atxn7l1
- Gene Name
- ataxin 7-like 1
View Full Product Details