Product Details

SNP ID
rs13481038
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:56040455 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTGTGCTCTCCAGCTTGGCTCGGAA[C/T]GGATGGACAATGGCCACGTAGCGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105244170 PubMed Links

Gene Details

Gene
LOC105244170
Gene Name
uncharacterized LOC105244170
There are no transcripts associated with this gene.

Gene
Nmur2
Gene Name
neuromedin U receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153079.4 533 Silent Mutation NP_694719.2

View Full Product Details