Product Details

SNP ID
rs13478607
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:5128915 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAATAGCATCTGTATCTCTAGGTAT[A/G]ATAGTTGATGCAATGCATCTCACAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ppp1r9a PubMed Links

Gene Details

Gene
Ppp1r9a
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 9A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181595.3 Intron NP_853626.1

View Full Product Details