Product Details

SNP ID
rs6243755
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:108737793 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAGAAAAAAAATCACAGACTTCCCA[C/T]TGTCCTCTGATTCATCTCTAATTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Syt1 PubMed Links

Gene Details

Gene
Syt1
Gene Name
synaptotagmin I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252341.1 Intron NP_001239270.1
NM_001252342.1 Intron NP_001239271.1
NM_009306.3 Intron NP_033332.1

View Full Product Details