Product Details
- SNP ID
-
rs13475824
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:40282099 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCCCAACCTAACCAGAAGCCCAG[C/T]GTAACATGGAATGGTGAAGAAGGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Il1r1
PubMed Links
Gene Details
- Gene
- Il1r1
- Gene Name
- interleukin 1 receptor, type I
View Full Product Details