Product Details

SNP ID
rs13475928
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:75525463 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACAGCCTGTGTGGCCTCCGTGGCAT[C/T]TACTCACAGCTGGAGTCCCTGGTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Stk11ip PubMed Links

Gene Details

Gene
Stk11ip
Gene Name
serine/threonine kinase 11 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_027886.3 526 Silent Mutation NP_082162.3

View Full Product Details