Product Details

SNP ID
rs4230463
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:70604117 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCTCTTAGCCACAGCTTTGAATAAG[T/C]CTTGGGTCTTGAGCAAGAGGTTTGA
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
Dmtn PubMed Links

Gene Details

Gene
Dmtn
Gene Name
dematin actin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252662.1 2196 UTR 3 NP_001239591.1
NM_001252663.1 2196 UTR 3 NP_001239592.1
NM_001252664.1 2196 UTR 3 NP_001239593.1
NM_001252665.1 2196 UTR 3 NP_001239594.1
NM_001252666.1 2196 UTR 3 NP_001239595.1
NM_013514.4 2196 UTR 3 NP_038542.1
Gene
Fam160b2
Gene Name
family with sequence similarity 160, member B2
There are no transcripts associated with this gene.

View Full Product Details