Product Details
- SNP ID
-
rs13481559
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:81655152 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATGTGGATGAAGGGCTCAGACTC[C/G]CAGAGGAACAGGAGGACACAACAGG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Ttc9
PubMed Links
Gene Details
- Gene
- Ttc9
- Gene Name
- tetratricopeptide repeat domain 9
View Full Product Details