Product Details
- SNP ID
-
rs3022854
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:170960884 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTCAACCATTGGGAACCAATCTC[G/T]TAGTGTCTGTTGGTTTCAGGACTCA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Fcgr2b
PubMed Links
Gene Details
- Gene
- Fcgr2b
- Gene Name
- Fc receptor, IgG, low affinity IIb
View Full Product Details