Product Details
- SNP ID
-
rs4223557
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:148404520 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTACATGTGACGTCATAAGCAAAG[G/T]CTTTGCTTGTGTTCTAGCGAAACTC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Sstr4
PubMed Links
Gene Details
- Gene
- Sstr4
- Gene Name
- somatostatin receptor 4
There are no transcripts associated with this gene.
- Gene
- Thbd
- Gene Name
- thrombomodulin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_009378.3 |
3669 |
UTR 3 |
|
|
NP_033404.1 |
View Full Product Details