Product Details

SNP ID
rs3723733
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:11454038 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGTTGATGGGATGCTTATTGGTTCA[G/A]CCACTATAGAAACCAATGTGGAGGG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
4930415F15Rik PubMed Links

Gene Details

Gene
4930415F15Rik
Gene Name
RIKEN cDNA 4930415F15 gene
There are no transcripts associated with this gene.

Gene
Zpbp
Gene Name
zona pellucida binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185153.1 Intron NP_001172082.1
NM_015785.2 Intron NP_056600.1

View Full Product Details