Product Details
- SNP ID
-
rs4174474
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:38174823 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAACTTTATGTAGTGCTGGGACTG[G/A]GCAGAATCACGCTAAGCTTAAATGT
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Gsk3b
PubMed Links
Gene Details
- Gene
- Gsk3b
- Gene Name
- glycogen synthase kinase 3 beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019827.6 |
|
Intron |
|
|
NP_062801.1 |
View Full Product Details