Product Details

SNP ID
rs3676196
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:42923810 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGGGTCCCACGAGACTGAAGAATTC[C/T]GTCACATTAAAACCAGGGGAAAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ppp2r2b PubMed Links

Gene Details

Gene
Ppp2r2b
Gene Name
protein phosphatase 2, regulatory subunit B, beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_028392.3 Intron NP_082668.1

View Full Product Details