Product Details

SNP ID
rs4222920
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:193272811 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGGGTGGCTGGGCCTGCAGAG[G/T]TAGCAGAGAGGAATGGGGTTGAGTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
G0s2 PubMed Links

Gene Details

Gene
G0s2
Gene Name
G0/G1 switch gene 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008059.3 Intron NP_032085.1
Gene
Hsd11b1
Gene Name
hydroxysteroid 11-beta dehydrogenase 1
There are no transcripts associated with this gene.

View Full Product Details