Product Details

SNP ID
rs4230683
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:30967015 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAACATTGTCTTTGTCTTTGCATTG[A/G]AGTAAGTCAGGAGGTGACATTGTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ctnnd2 PubMed Links

Gene Details

Gene
Ctnnd2
Gene Name
catenin (cadherin associated protein), delta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008729.2 Intron NP_032755.2

View Full Product Details