Product Details
- SNP ID
-
rs6188239
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:34331233 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTGCCCAATAGTTTTCCTTTTGA[A/G]TAATTACTGTCCCCATGTGTTGAGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Matn2
PubMed Links
Gene Details
- Gene
- Matn2
- Gene Name
- matrilin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016762.2 |
|
Intron |
|
|
NP_058042.2 |
View Full Product Details