Product Details

SNP ID
rs4230941
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:89131096 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCCAAGGTGCCGTGAAGCTCC[C/G]AGCCAACCTCCTTTGGATACCCCCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Hdac10 PubMed Links

Gene Details

Gene
Hdac10
Gene Name
histone deacetylase 10
There are no transcripts associated with this gene.

Gene
Mapk12
Gene Name
mitogen-activated protein kinase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013871.3 1378 UTR 3 NP_038899.1
Gene
Tubgcp6
Gene Name
tubulin, gamma complex associated protein 6
There are no transcripts associated with this gene.

View Full Product Details