Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDSN
Entrez Gene ID
102118455
Gene Name
corneodesmosin
Gene Aliases
-
Chromosome Location
Chr.4: 139851476 - 139858087 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005553588.2 XP_005553645.1 1 - 2 1852 106

View Full Product Details