Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAMD13
Entrez Gene ID
102118637
Gene Name
sterile alpha motif domain containing 13
Gene Aliases
-
Chromosome Location
Chr.1: 142906590 - 142957139 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005542892.2 XP_005542949.1 1 - 2 97 59

View Full Product Details