Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LHFPL1
Entrez Gene ID
102117018
Gene Name
LHFPL tetraspan subfamily member 1
Gene Aliases
-
Chromosome Location
Chr.X: 109683832 - 109745875 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005594401.2 XP_005594458.1 1 - 2 388 67

View Full Product Details