Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXCL13
Entrez Gene ID
102123271
Gene Name
C-X-C motif chemokine ligand 13
Gene Aliases
-
Chromosome Location
Chr.5: 56870599 - 56875595 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284543.1 NP_001271472.1 1 - 2 108 75
GenBank mRNA JQ288995.1 - 1 - 2 108 75

View Full Product Details