Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBED3
Entrez Gene ID
102141982
Gene Name
zinc finger BED-type containing 3
Gene Aliases
-
Chromosome Location
Chr.6: 75258423 - 75271475 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005557219.2 XP_005557276.1 2 - 3 338 99

View Full Product Details