Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDN2
Entrez Gene ID
102134258
Gene Name
endothelin 2
Gene Aliases
ET-2, PPET2
Chromosome Location
Chr.1: 186216134 - 186222114 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005543767.2 XP_005543824.2 3 - 4 481 138
GenBank mRNA U20580.1 - 1 - 2 126 138

View Full Product Details