Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFF1
Entrez Gene ID
102127360
Gene Name
trefoil factor 1
Gene Aliases
-
Chromosome Location
Chr.3: 4402450 - 4407162 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005548621.1 XP_005548678.1 2 - 3 487 184

View Full Product Details