Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSEN15
Entrez Gene ID
102127302
Gene Name
tRNA splicing endonuclease subunit 15
Gene Aliases
-
Chromosome Location
Chr.1: 42016001 - 42038671 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005540221.2 XP_005540278.1 4 - 5 588 131

View Full Product Details