Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTR3C
Entrez Gene ID
102122102
Gene Name
5-hydroxytryptamine receptor 3C
Gene Aliases
-
Chromosome Location
Chr.2: 88723109 - 88730973 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005548435.1 XP_005548492.1 2 - 3 233 117

View Full Product Details