Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRYGB
Entrez Gene ID
102146176
Gene Name
crystallin gamma B
Gene Aliases
-
Chromosome Location
Chr.12: 97764044 - 97768624 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005574097.2 XP_005574154.1 1 - 2 449 84

View Full Product Details