Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STOX1
Entrez Gene ID
102116958
Gene Name
storkhead box 1
Gene Aliases
-
Chromosome Location
Chr.9: 66907177 - 66917861 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005565679.2 XP_005565736.1 2 - 3 2928 169

View Full Product Details