Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXHXorf58
Entrez Gene ID
102115232
Gene Name
chromosome X open reading frame, human CXorf58
Gene Aliases
-
Chromosome Location
Chr.X: 21872633 - 21906644 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005593180.2 XP_005593237.1 8 - 9 1282 144

View Full Product Details